5-Methyl-2-hexanone ≥98.0%

Supplier: TCI America
Danger

Synonyms: Isoamyl methyl ketone, Methyl isoamyl ketone

I0087-500ML I0087-25ML
TCI0087-500MLEA 38 USD
TCI0087-500ML TCI0087-25ML
5-Methyl-2-hexanone ≥98.0%
5-Methyl-2-hexanone

CAS Number: 110-12-3
MDL Number: MFCD00008950
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Melting point (°C): -74
Flash Point (°C): 36
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: 5-Methyl-2-hexanone

Formula: C₇H₁₄O
Boiling Pt: 144 °C
Melting Pt: –74 °C
Density: 0.81 (20 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00008950
CAS Number: 110-12-3
EINECS: 203-737-8
UN: 2302
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cf0b6d70c679f5901d6cc74ff38e3e9889ad641&vid=478a9be903db5124bc507fb2a29f65e402327706&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR