Diisoamyl ether (mixture of isomers) ≥98.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with Stabilised

I0086-500ML I0086-25ML
TCI0086-500MLEA 112 USD
TCI0086-500ML TCI0086-025ML
Diisoamyl ether (mixture of isomers) ≥98.0% stabilized
Diisoamyl ether (mixture of isomers)

(stabilized with BHT)
CAS Number: 544-01-4
MDL Number: MFCD00008947
Molecular Formula: C10H22O
Molecular Weight: 158.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Flash Point (°C): 48
Specific Gravity (20/20): 0.78


Other Product Names / Synonyms: Diisoamyl Ether, Isopentyl Ether, Diisopentyl Ether

Formula: C₁₀H₂₂O
MW: 158.28 g/mol
Boiling Pt: 173 °C
Density: 0.78 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00008947
CAS Number: 544-01-4
EINECS: 208-857-4
UN: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62fb20b3780f2691b55992319ac34d8b7111ff8e&vid=62fb20b3780f2691b55992319ac34d8b7111ff8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR