Isopentylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Amino-3-methylbutane, Isopentyl amine, Isoamylamine, 3-Methylbutylamine, Leucamine

I0082-500ML I0082-25ML I0082-100ML
TCI0082-500MLEA 233 USD
TCI0082-500ML TCI0082-25ML TCI0082-100ML
Isopentylamine ≥98.0% (by GC, titration analysis)
Isopentylamine

CAS Number: 107-85-7
MDL Number: MFCD00008203
Molecular Formula: C5H13N
Molecular Weight: 87.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): -1
Specific Gravity (20/20): 0.75


Other Product Names / Synonyms: 1-Amino-3-methylbutane, Isopentylamine, 3-Methylbutylamine

Formula: C₅H₁₃N
MW: 87.16 g/mol
Boiling Pt: 97 °C
Melting Pt: –68 °C
Density: 0.75 (20 °C)
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008203
CAS Number: 107-85-7
EINECS: 203-526-0
UN: 1106
ADR: 3,II
Merck Index: 13,05128
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65a9477bf00f499ebc6d894aee427dcd49e04f98&vid=faae898fa059b780c09c81af8dcb70db4d0fee2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR