2-Iodopropane ≥99.0% stabilized

Supplier: TCI America
Danger

Synonyms: Isopropyl iodide

Stabilization: Stabilised with Stabilised

I0069-25G I0069-100G
TCI0069-25GEA 48.56 USD
TCI0069-25G TCI0069-100G
2-Iodopropane ≥99.0% stabilized
2-Iodopropane

(stabilized with Copper chip)
CAS Number: 75-30-9
MDL Number: MFCD00001071
Molecular Formula: C3H7I
Molecular Weight: 169.99
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Melting point (°C): -90
Flash Point (°C): 42
Specific Gravity (20/20): 1.71


Other Product Names / Synonyms: Isopropyl Iodide

Formula: C₃H₇I
MW: 169.99 g/mol
Boiling Pt: 90 °C
Melting Pt: –90 °C
Density: 1.71 (20 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00001071
CAS Number: 75-30-9
EINECS: 200-859-3
UN: 2392
ADR: 3,III
Merck Index: 13,05234
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec788505b6846ad10a477ebbf13f4c3d745af374&vid=d917a9e77fbba469f538fd38c0a44b97e8cbe9ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR