1-Iodopropane ≥98.0% stabilized

Supplier: TCI America
Danger

Synonyms: 1-Propyl iodide

Stabilization: Stabilised with Stabilised

I0068-500G I0068-25G I0068-100G
TCI0068-500GEA 569 USD
TCI0068-500G TCI0068-25G TCI0068-100G
1-Iodopropane ≥98.0% stabilized
1-Iodopropane

(stabilized with Copper chip)
CAS Number: 107-08-4
MDL Number: MFCD00001095
Molecular Formula: C3H7I
Molecular Weight: 169.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 44
Freezing point (°C): -101
Specific Gravity (20/20): 1.75


Other Product Names / Synonyms: Propyl Iodide

Formula: C₃H₇I
MW: 169.99 g/mol
Boiling Pt: 102 °C
Melting Pt: –98 °C
Density: 1.75 (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00001095
CAS Number: 107-08-4
EINECS: 203-460-2
UN: 2392
ADR: 3,III
Merck Index: 12,08047
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2355784bd416f7570ee096c5fbb9eb84620cb24f&vid=33df6068363349f90fc01f705038506bcf671ab7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR