1-Iodobutane ≥98.0% stabilized

Supplier: TCI America
Danger

Synonyms: 1-Butyl iodide

Stabilization: Stabilised with Stabilised

I0055-500G I0055-25G
TCI0055-500GEA 242 USD
TCI0055-500G TCI0055-25G
1-Iodobutane ≥98.0% stabilized
1-Iodobutane

(stabilized with Copper chip)
CAS Number: 542-69-8
MDL Number: MFCD00001098
Molecular Formula: C4H9I
Molecular Weight: 184.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 49
Specific Gravity (20/20): 1.62


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Butyl Iodide

Formula: C₄H₉I
MW: 184.02 g/mol
Boiling Pt: 130 °C
Melting Pt: –103 °C
Density: 1.62 (20 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00001098
CAS Number: 542-69-8
EINECS: 208-824-4
UN: 1993
ADR: 3,III
Merck Index: 13,01572
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=723ae0732ef9a14c17dda4d1534ab6a3284ab08b&vid=e423323ae1d40e0e7e3db0b6024898411a874fd9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR