3-Hydrazinobenzoic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Hydrazinylbenzoic acid

H0799-5G H0799-25G
TCH0799-005GEA 43.9 USD
TCH0799-005G TCH0799-025G
3-Hydrazinobenzoic acid ≥97.0% (by titrimetric analysis)
3-Hydrazinobenzoic acid

CAS Number: 38235-71-1
MDL Number: MFCD00191447
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 183

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 183 °C
MDL Number: MFCD00191447
CAS Number: 38235-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b89a0c6314c4c18807eddf6a5d8c8e08355a0483&vid=b89a0c6314c4c18807eddf6a5d8c8e08355a0483&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR