N-(Hydroxymethyl)phthalimide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2-(Hydroxymethyl)isoindoline-1,3-dione

H0648-250G H0648-25G
TCH0648-250GEA 152.63 USD
TCH0648-250G TCH0648-025G
N-(Hydroxymethyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
N-(Hydroxymethyl)phthalimide

CAS Number: 118-29-6
MDL Number: MFCD00005899
Molecular Formula: C9H7NO3
Molecular Weight: 177.16
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 147

Formula: C₉H₇NO₃
MW: 177.16 g/mol
Boiling Pt: 415 °C (1013 hPa)
Melting Pt: 147 °C
MDL Number: MFCD00005899
CAS Number: 118-29-6
EINECS: 204-241-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7cfbf338b0579c54da3ba256bf341cdb678e4b5&vid=e7cfbf338b0579c54da3ba256bf341cdb678e4b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR