Ethyl heptafluorobutyrate ≥97.0%

Supplier: TCI America

Synonyms: Ethyl 2,2,3,3,4,4,4-heptafluorobutanoate, Ethyl perfluorobutyrate

H0594-5G H0594-25G
TCH0594-005GEA 80 USD
TCH0594-005G TCH0594-025G
Ethyl heptafluorobutyrate ≥97.0%
Ethyl heptafluorobutyrate

CAS Number: 356-27-4
MDL Number: MFCD00000434
Molecular Formula: C6H5F7O2
Molecular Weight: 242.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 14
Specific Gravity (20/20): 1.40


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Heptafluorobutyric Acid Ethyl Ester, Perfluorobutyric Acid Ethyl Ester, Ethyl Perfluorobutyrate

Formula: C₆H₅F₇O₂
MW: 242.09 g/mol
Boiling Pt: 96 °C
Density: 1.40 (20 °C)
Flash Pt: 14 °C
MDL Number: MFCD00000434
CAS Number: 356-27-4
EINECS: 206-602-1
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4177ec33b2993964cb20171cf745bdc005a8d0a2&vid=4177ec33b2993964cb20171cf745bdc005a8d0a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR