α,α,α,α',α',α'-Hexafluoro-m-xylene ≥98.0%

Supplier: TCI America
Danger

Synonyms: 1,3-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-m-xylene

H0585-500G H0585-25G H0585-100G
TCH0585-500GEA 170.87 USD
TCH0585-500G TCH0585-25G TCH0585-100G
α,α,α,α',α',α'-Hexafluoro-m-xylene ≥98.0%
α,α,α,α',α',α'-Hexafluoro-m-xylene

CAS Number: 402-31-3
MDL Number: MFCD00000392
Molecular Formula: C8H4F6
Molecular Weight: 214.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 26
Specific Gravity (20/20): 1.40


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,alpha',alpha',alpha'-Hexafluoro-m-xylene

Formula: C₈H₄F₆
MW: 214.11 g/mol
Boiling Pt: 116 °C
Melting Pt: –35 °C
Density: 1.40 (20 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00000392
CAS Number: 402-31-3
EINECS: 206-939-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e19c77f23b3f581b2366df7d6a4b7116968fb284&vid=b87397f93be6a056cb6e7765e50dff5367964d82&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR