trans-3-Hexene ≥99.0%
Supplier: TCI America
H0399-5ML
H0399-1ML
TCH0399-005MLEA
132.16
USD
TCH0399-005ML
TCH0399-1ML
trans-3-Hexene ≥99.0%
trans-3-Hexene
CAS Number: 13269-52-8
MDL Number: MFCD00009386
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): -12
Specific Gravity (20/20): 0.68
Formula:
C₆H₁₂ Boiling Pt: 67 °C Melting Pt: –115.4 °C Density: 0.68 (20 °C) Flash Pt: –12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009386 CAS Number: 13269-52-8 EINECS: 236-261-4 UN: 3295 ADR: 3,II |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...