trans-2-Hexenal ≥97.0%

Supplier: TCI America

Synonyms: (E)-2-Hexenal, trans-2-Hexenal

H0345-25ML H0345-100ML
TCH0345-25MLEA 26 USD
TCH0345-25ML TCH0345-100ML
trans-2-Hexenal ≥97.0%
trans-2-Hexenal

CAS Number: 6728-26-3
MDL Number: MFCD00007008
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 43
Specific Gravity (20/20): 0.85
Storage Temperature: 0-10°C


Product Categories: Analytical Science, Odor Deterioration Factors of Cosmetics
Other Product Names / Synonyms: trans-2-Hexenyl Aldehyde, Leaf Aldehyde, trans-3-Propylacrolein

Formula: C₆H₁₀O
Boiling Pt: 146 °C
Density: 0.85 (20 °C)
Flash Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007008
CAS Number: 6728-26-3
EINECS: 229-778-1
UN: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71678219c2ca5a9177b78b5eac4d85ab62200edb&vid=71bf61ca4ec808310e7876b2e646d2bd4942e102&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR