2-Hydroxypyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
H0330-250G
H0330-25G
TCH0330-250GEA
362
USD
TCH0330-250G
TCH0330-025G
2-Hydroxypyridine ≥98.0% (by GC, titration analysis)
2-Hydroxypyridine
CAS Number: 142-08-5
MDL Number: MFCD00006268
Molecular Formula: C5H5NO
Molecular Weight: 95.10
Purity/Analysis Method:
Other Product Names / Synonyms: 2-Pyridinol, 2-Pyridone
Formula:
C₅H₅NO MW: 95.10 g/mol Boiling Pt: 281 °C Melting Pt: 107 °C Density: 1.085 g/cm³ (30 °C) Flash Pt: 161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006268 CAS Number: 142-08-5 EINECS: 205-520-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...