1,6-Diisocyanatohexane ≥98.0%

Supplier: TCI America
Danger

H0324-25ML H0324-100ML
TCH0324-25MLEA 29.48 USD
TCH0324-25ML TCH0324-100ML
1,6-Diisocyanatohexane ≥98.0%
1,6-Diisocyanatohexane

CAS Number: 822-06-0
MDL Number: MFCD00002047
Molecular Formula: C8H12N2O2
Molecular Weight: 168.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 255
Flash Point (°C): 138
Specific Gravity (20/20): 1.05
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,6-Diisocyanatohexane

Formula: C₈H₁₂N₂O₂
Boiling Pt: 255 °C
Melting Pt: –67 °C
Density: 1.05 (20 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00002047
CAS Number: 822-06-0
EINECS: 212-485-8
UN: 2281
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f64a050b16dcbfe2c9afb36e0e45269e6d2e6f9b&vid=f64a050b16dcbfe2c9afb36e0e45269e6d2e6f9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR