Oxybenzone ≥99.0%

Supplier: TCI America

Synonyms: Eusolex® 4360, 2-Hydroxy-4-methoxybenzophenone, (2-Hydroxy-4-methoxyphenyl)(phenyl)methanone

H0266-500G H0266-25G
TCH0266-500GEA 122 USD
TCH0266-500G TCH0266-25G
Oxybenzone ≥99.0%
Oxybenzone

CAS Number: 131-57-7
MDL Number: MFCD00008387
Molecular Formula: C14H12O3
Molecular Weight: 228.25
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 63
Flash Point (°C): 188


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Oxybenzone

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Boiling Pt: 370 °C (1013 hPa)
Melting Pt: 63 °C
Density: 1.3397 g/cm³ (25 °C)
Flash Pt: 188 °C
MDL Number: MFCD00008387
CAS Number: 131-57-7
EINECS: 205-031-5
Merck Index: 12,07088
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=deace785d2da5249aa1e6fae6d402108801714cb&vid=deace785d2da5249aa1e6fae6d402108801714cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR