2-Hydroxy-m-anisaldehyde ≥99.0%

Supplier: TCI America

Synonyms: o-Vanillin, 2-Hydroxy-3-methoxybenzaldehyde

H0262-25G H0262-500G H0262-100G
TCH0262-025GEA 43.9 USD
TCH0262-025G TCH0262-500G TCH0262-100G
2-Hydroxy-m-anisaldehyde ≥99.0%
2-Hydroxy-m-anisaldehyde

CAS Number: 148-53-8
MDL Number: MFCD00003322
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 268
Melting point (°C): 44
Flash Point (°C): 110


Other Product Names / Synonyms: 2-Hydroxy-3-methoxybenzaldehyde, 3-Methoxysalicylaldehyde

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 268 °C
Melting Pt: 44 °C
Density: 1.2039 g/cm³ (45 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003322
CAS Number: 148-53-8
EINECS: 205-715-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee811599c2603ce1b901ddbceceaec385739ff96&vid=05f4aa6dd5114599b9e45d1858b8fbd629ef960f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR