4-Heptyloxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-(Heptyloxy)benzoic acid

H0248-500G H0248-25G
TCH0248-500GEA 837 USD
TCH0248-500G TCH0248-025G
4-Heptyloxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Heptyloxybenzoic acid

CAS Number: 15872-42-1
MDL Number: MFCD00013992
Molecular Formula: C14H20O3
Molecular Weight: 236.31
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 145

Formula: C₁₄H₂₀O₃
MW: 236.31 g/mol
Boiling Pt: 362 °C (1013 hPa)
Melting Pt: 145 °C
Density: 1.183 g/cm³ (20 °C)
MDL Number: MFCD00013992
CAS Number: 15872-42-1
EINECS: 239-995-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc9a9b21beed3962ea56ad1554306cf747027d4e&vid=17586798a91f2378df2c2091d649957ab0e3507a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR