Acetoin ≥98.0% (May exist as crystalline dimer)

Supplier: TCI America

Synonyms: Acetyl methyl carbinol, 3-Hydroxybutanone, 3-Hydroxy-2-butanone

H0225-500G H0225-25G
TCH0225-500GEA 573 USD
TCH0225-500G TCH0225-25G
Acetoin ≥98.0% (May exist as crystalline dimer)
Acetoin

CAS Number: 513-86-0
MDL Number: MFCD00004521
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 144
Flash Point (°C): 46
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Acetylmethylcarbinol, 3-Hydroxy-2-butanone

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 144 °C
Melting Pt: 20 °C
Density: 0.998 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038696
CAS Number: 513-86-0
EINECS: 208-174-1
UN: 2621
ADR: 3,III
Merck Index: 13,00065
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ec11d2db1ea00b7885a9d51a04d2f25285ae314&vid=e9029a8ae14be8319ff9e0896ff914d6328edeb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR