4-Hydroxybenzaldehyde ≥98.0%

Supplier: TCI America
Warning

H0198-25G H0198-500G H0198-100G
TCH0198-025GEA 19.31 USD
TCH0198-025G TCH0198-500G TCH0198-100G
4-Hydroxybenzaldehyde ≥98.0%
4-Hydroxybenzaldehyde

CAS Number: 123-08-0
MDL Number: MFCD00006939
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 117
Flash Point (°C): 78

Formula: C₇H₆O₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00006939
CAS Number: 123-08-0
EINECS: 204-599-1
Merck Index: 13,04836
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b9db03c9466f34fa7d53422acb97dd8475a3245&vid=c7656203f09518b5537188ef468b1c75b8cdec60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR