3-Hydroxybenzaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

H0197-25G H0197-500G
TCH0197-025GEA 67 USD
TCH0197-025G TCH0197-500G
3-Hydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
3-Hydroxybenzaldehyde

CAS Number: 100-83-4
MDL Number: MFCD00003368
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 168
Melting point (°C): 104

Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 168 °C
Melting Pt: 104 °C
Density: 1.1299 g/cm³ (114 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003368
CAS Number: 100-83-4
EINECS: 202-892-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fadca81c1ddd2d1ba6c393feec799617e8fe2fce&vid=40a421125ff8a3caa135769cb9bda83706de0b56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR