Hydroquinone ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1,4-Dihydroxybenzene, Quinol
H0186-500G
H0186-25G
TCH0186-500GEA
43.9
USD
TCH0186-500G
TCH0186-25G
Hydroquinone ≥99.0%
Hydroquinone
CAS Number: 123-31-9
MDL Number: MFCD00002339
Molecular Formula: C6H6O2
Molecular Weight: 110.11
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 282
Melting point (°C): 173
Flash Point (°C): 165
Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Polymerization Inhibitors
Other Product Names / Synonyms: 1,4-Dihydroxybenzene, Quinol, HQ
Formula:
C₆H₄(OH)₂ Boiling Pt: 282 °C Melting Pt: 173 °C Density: 1.332 g/cm³ (20 °C) Flash Pt: 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002339 CAS Number: 123-31-9 EINECS: 204-617-8 UN: 3077 ADR: 9,III Merck Index: 13,04833 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...