Hydrazinium monobromide ≥98.0%
Supplier: TCI America
Synonyms:
Hydrazine monohydrobromide
H0173-500G
H0173-25G
TCH0173-500GEA
289
USD
TCH0173-500G
TCH0173-25G
Hydrazinium monobromide ≥98.0%
Hydrazinium monobromide
CAS Number: 13775-80-9
MDL Number: MFCD00065394
Molecular Formula: H4N2
Molecular Weight: 112.96
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 88
Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: Hydrazinium Monobromide
Formula:
H₅BrN₂ Melting Pt: 88 °C |
MDL Number:
MFCD00065394 CAS Number: 13775-80-9 EINECS: 237-412-7 UN: 2928 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...