Hydrazinium hydroxide ≥98% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Hydrazine hydrate, Hydrazine monohydrate

H0172-25ML H0172-500ML
TCH0172-25MLEA 29.35 USD
TCH0172-25ML TCH0172-500ML
Hydrazinium hydroxide ≥98% (by titrimetric analysis)
Hydrazinium hydroxide

CAS Number: 7803-57-8 MDL Number: MFCD00149931 Molecular Formula: H4N2 Molecular Weight: 32.05 Purity/Analysis Method: 98.0% (T) Form: Clear Liquid Boiling point (°C): 119 Melting point (°C): -52 Flash Point (°C): 75 Specific Gravity (20/20): 1.03

Formula: NH₂NH₂·H₂O
MW: 50.06 g/mol
Boiling Pt: 119 °C
Melting Pt: -52 °C
Density: 1.03 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00149931
CAS Number: 7803-57-8
EINECS: 206-114-9
UN: 2030
ADR: 8,II
Merck Index: 13,04790
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1a118e8bc0b119eedd1ad089f1b7216dd82c3e2&vid=b1a118e8bc0b119eedd1ad089f1b7216dd82c3e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR