Hydrazine dihydrochloride ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Hydrazine Dihydrochloride, Hydrazinium dichloride
H0170-500G
H0170-25G
TCH0170-500GEA
99.14
USD
TCH0170-500G
TCH0170-25G
Hydrazine dihydrochloride ≥98.0%
Hydrazine dihydrochloride
CAS Number: 5341-61-7
MDL Number: MFCD00064543
Molecular Formula: H4N2
Molecular Weight: 104.96
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 200
Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: Hydrazinium Dichloride
Formula:
H₆Cl₂N₂ MW: 104.97 g/mol Melting Pt: 189 Density: 1.42 g/cm³ (20 °C) |
MDL Number:
MFCD00064543 CAS Number: 5341-61-7 EINECS: 226-283-2 UN: 2928 ADR: 6.1 II Merck Index: 13,04789 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...