Hexylamine ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 1-Hexylamine, 1-Aminohexane

H0134-500ML H0134-25ML H0134-100ML
TCH0134-500MLEA 136.3 USD
TCH0134-500ML TCH0134-25ML TCH0134-100ML
Hexylamine ≥99.0% (by GC, titration analysis)
Hexylamine

CAS Number: 111-26-2
MDL Number: MFCD00008240
Molecular Formula: C6H15N
Molecular Weight: 101.19
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 130
Melting point (°C): -19
Flash Point (°C): 27
Specific Gravity (20/20): 0.77


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Aminohexane

Formula: H₃C(CH₂)₄CH₂NH₂
MW: 101.19 g/mol
Boiling Pt: 130 °C
Melting Pt: -19 °C
Density: 0.77 (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00008240
CAS Number: 111-26-2
EINECS: 203-851-8
UN: 2734
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b8279b00ac09bbf6259ae7e2bc1e9223f0269d8&vid=36e4f259f35c6de7d206ad3b7a5e399ad03b7654&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR