(±)-2-Hexanol ≥98.0%

Supplier: TCI America

H0131-25ML H0131-100ML
TCH0131-25MLEA 49.12 USD
TCH0131-25ML TCH0131-100ML
(±)-2-Hexanol ≥98.0%
(±)-2-Hexanol

CAS Number: 626-93-7
MDL Number: MFCD00004585
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 139
Flash Point (°C): 41
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: 2-Hexyl Alcohol

Formula: C₆H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00004585
CAS Number: 626-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a6e95f48f5a8567d32c0e14217905109f543637&vid=30d09f2f591bc9aa534924eb9b26d59e117b13b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR