Ethyl hexanoate ≥99.0%

Supplier: TCI America

Synonyms: Ethyl caproate, Caproic acid ethyl ester, Hexanoic acid ethyl ester

H0108-500ML H0108-25ML
TCH0108-500MLEA 59.02 USD
TCH0108-500ML TCH0108-025ML
Ethyl hexanoate ≥99.0%
Ethyl hexanoate

CAS Number: 123-66-0
MDL Number: MFCD00009511
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 167
Flash Point (°C): 55
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Ethyl Caproate, Hexanoic Acid Ethyl Ester

Formula: C₈H₁₆O₂
Boiling Pt: 167 °C
Melting Pt: -67 °C
Density: 0.87 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00009511
CAS Number: 123-66-0
EINECS: 204-640-3
UN: 3272
ADR: 3,III
Merck Index: 13,03812
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f281167feb22a4941bd121c0fea959b0c862c36&vid=6f281167feb22a4941bd121c0fea959b0c862c36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR