Caproic acid ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Hexanoic acid
H0105-500ML
H0105-25ML
TCH0105-500MLEA
35.6
USD
TCH0105-500ML
TCH0105-25ML
Caproic acid ≥98.0%
Caproic acid
CAS Number: 142-62-1
MDL Number: MFCD00004421
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 205
Melting point (°C): -3
Flash Point (°C): 108
Specific Gravity (20/20): 0.93
Formula:
C₆H₁₂O₂ Boiling Pt: 205 °C Melting Pt: –3 °C Density: 0.93 (20 °C) Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004421 CAS Number: 142-62-1 EINECS: 205-550-7 UN: 2829 ADR: 8,III Merck Index: 13,01765 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...