1,5-Hexadiene ≥97.0%

Supplier: TCI America

Synonyms: Hexa-1,5-diene

H0084-25ML H0084-100ML
TCH0084-25MLEA 24 USD
TCH0084-25ML TCH0084-100ML
1,5-Hexadiene ≥97.0%
1,5-Hexadiene

CAS Number: 592-42-7
MDL Number: MFCD00008666
Molecular Formula: C6H10
Molecular Weight: 82.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 59
Flash Point (°C): -46
Freezing point (°C): -141
Specific Gravity (20/20): 0.69


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Monomers
Other Product Names / Synonyms: Biallyl, Diallyl

Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 59 °C
Melting Pt: -141 °C
Density: 0.69 (20 °C)
Flash Pt: -46 °C
Storage Temperature: Ambient
MDL Number: MFCD00008666
CAS Number: 592-42-7
EINECS: 209-754-7
UN: 2458
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95415d12e4e916313cc90ab74a6be87910546cfa&vid=95415d12e4e916313cc90ab74a6be87910546cfa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR