Cetrimonium chloride ≥95.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
CAS Number: 112-02-7
MDL Number: MFCD00011773
Molecular Formula: C19H42ClN
Molecular Weight: 320.00
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 234
Product Categories: Materials Chemistry, Cationic Surfactants, Bioscience, Quaternary Ammonium Salts for Clinical Chemistry
Other Product Names / Synonyms: Cetyltrimethylammonium Chloride, HTAC, CTAC
Formula:
C₁₉H₄₂ClN Melting Pt: 234 °C Density: 0.9 g/cm³ (20 °C) |
MDL Number:
MFCD00011773 CAS Number: 112-02-7 EINECS: 203-928-6 UN: 3077 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...