Hexadecylamine ≥95.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1-Hexadecylamine, n-Hexadecylamine, 1-Aminohexadecane
H0074-500G
H0074-25G
TCH0074-500GEA
88.23
USD
TCH0074-500G
TCH0074-25G
Hexadecylamine ≥95.0%
Hexadecylamine
CAS Number: 143-27-1
MDL Number: MFCD00008158
Molecular Formula: C16H35N
Molecular Weight: 241.46
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 330
Flash Point (°C): 137
Freezing point (°C): 44
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Aminohexadecane, Cetylamine
Formula:
H₃C(CH₂)₁₄CH₂NH₂ MW: 241.46 g/mol Boiling Pt: 330 °C Melting Pt: 43…46 °C Density: 0.7821 g/cm³ (60 °C) Flash Pt: 137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008158 CAS Number: 143-27-1 EINECS: 205-596-8 UN: 3259 ADR: 8 II |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...