1-Heptene ≥98.0%
Supplier: TCI America
|
Danger
|
H0042-25ML
H0042-100ML
TCH0042-25MLEA
99.62
USD
TCH0042-25ML
TCH0042-100ML
1-Heptene ≥98.0%
1-Heptene
CAS Number: 592-76-7
MDL Number: MFCD00009531
Molecular Formula: C7H14
Molecular Weight: 98.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): -4
Specific Gravity (20/20): 0.70
Formula:
C₇H₁₄ MW: 98.19 g/mol Boiling Pt: 94 °C Melting Pt: –119 °C Density: 0.70 (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009531 CAS Number: 592-76-7 EINECS: 209-767-8 UN: 2278 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...