Heptanoyl chloride ≥98.0%

Supplier: TCI America

Synonyms: Heptanoylchloride

H0040-25G H0040-500G H0040-100G
TCH0040-025GEA 41.69 USD
TCH0040-025G TCH0040-500G TCH0040-100G
Heptanoyl chloride ≥98.0%
Heptanoyl chloride

CAS Number: 2528-61-2
MDL Number: MFCD00000763
Molecular Formula: C7H13ClO
Molecular Weight: 148.63
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 58
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Enanthyl Chloride

Formula: C₇H₁₃ClO
Boiling Pt: 175 °C
Melting Pt: –83.8 °C
Density: 0.96 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00000763
CAS Number: 2528-61-2
EINECS: 219-775-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35c11222465d50041f7b06af58454ecdbc0ee470&vid=35c11222465d50041f7b06af58454ecdbc0ee470&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR