2-Heptanone ≥98.0%

Supplier: TCI America
Danger

H0037-500ML H0037-25ML
TCH0037-500MLEA 93.11 USD
TCH0037-500ML TCH0037-25ML
2-Heptanone ≥98.0%
2-Heptanone

CAS Number: 110-43-0
MDL Number: MFCD00009513
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Melting point (°C): -36
Flash Point (°C): 42
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Amyl Methyl Ketone, Methyl Pentyl Ketone

Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 151 °C
Melting Pt: –36 °C
Density: 0.82 (20 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00009513
CAS Number: 110-43-0
EINECS: 203-767-1
UN: 1110
ADR: 3,III
Merck Index: 13,04683
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e55993e592e22708c9e92618b34d4ea3fa8dc539&vid=d1c3c2c1ee35bcacd958d49a29fcb6e8179ea6de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR