Heptanoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Heptylic acid, Enanthic acid

H0030-500ML H0030-25ML
TCH0030-500MLEA 50.86 USD
TCH0030-500ML TCH0030-25ML
Heptanoic acid ≥98.0% (by titrimetric analysis)
Heptanoic acid

CAS Number: 111-14-8
MDL Number: MFCD00004426
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 223
Melting point (°C): -8
Flash Point (°C): 120
Specific Gravity (20/20): 0.92


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Lipids
Other Product Names / Synonyms: Enanthic Acid

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 223 °C
Melting Pt: –8 °C
Density: 0.92 (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00004426
CAS Number: 111-14-8
EINECS: 203-838-7
UN: 3265
ADR: 8,III
Merck Index: 13,04680
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c398457150057d694b2c2c6eb880e799a7f155a&vid=d1511ebd9c1342c1d875d94749293108c53b3ca4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR