Margaric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

H0019-500G H0019-25G H0019-100G
TCH0019-500GEA 938.4 USD
TCH0019-500G TCH0019-025G TCH0019-100G
Margaric acid ≥98.0% (by GC, titration analysis)
Margaric acid

CAS Number: 506-12-7
MDL Number: MFCD00002751
Molecular Formula: C17H34O2
Molecular Weight: 270.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 61


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Margaric Acid

Formula: C₁₇H₃₄O₂
Boiling Pt: 220 °C
Melting Pt: 61 °C
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002751
CAS Number: 506-12-7
EINECS: 208-027-1
Merck Index: 13,05775
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9a7ed9d517e3934bd5d8bc729ea44912cfbb41d&vid=b53d67b952755420f33b8d00ba1befd114ed0025&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR