2,5-Piperazinedione ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
Glycine anhydride, Piperazine-2,5-dione
G0100-500G
G0100-25G
TCG0100-500GEA
747
USD
TCG0100-500G
TCG0100-025G
2,5-Piperazinedione ≥98.0% (by total nitrogen basis)
2,5-Piperazinedione
CAS Number: 106-57-0
MDL Number: MFCD00006009
Molecular Formula: C4H6N2O2
Molecular Weight: 114.10
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Other Product Names / Synonyms: 2,5-Diketopiperazine, 2,5-Dioxopiperazine, Glycocoll Anhydride, 2,5-Piperazinedione
Formula:
C₄H₆N₂O₂ MW: 114.1 g/mol |
MDL Number:
MFCD00006009 CAS Number: 106-57-0 EINECS: 203-411-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...