Glutaraldehyde ∼50% ∼5.6 mol/L in water

Supplier: TCI America
Danger

Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial

G0068-500ML G0068-25ML
TCG0068-500MLEA 48.1 USD
TCG0068-500ML TCG0068-25ML
Glutaraldehyde ∼50% ∼5.6 mol/L in water
Glutaraldehyde

CAS Number: 111-30-8
MDL Number: MFCD00007025
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.13

Formula: C₅H₈O₂
MW: 100.12
Boiling Pt: 188
Melting Pt: -10
Density: 1.13 (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2927
ADR: 6.1 II
Merck Index: 13,04485
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=319c6f1d87cbbe3515edacd7047ed2492c554d90&vid=954ae2e5dd5c47b868a670c765c06816082bcbbd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR