L(+)-Glutamic acid hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(S)-2-Aminopentanedioic acid hydrochloride, 2-Aminoglutaric acid hydrochloride
G0060-25G
G0060-500G
TCG0060-025GEA
21.18
USD
TCG0060-025G
TCG0060-500G
L(+)-Glutamic acid hydrochloride ≥98.0% (by titrimetric analysis)
L(+)-Glutamic acid hydrochloride
CAS Number: 138-15-8
MDL Number: MFCD00012619
Molecular Formula: C5H9NO4
Molecular Weight: 183.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 24 deg (C=6, H2O)
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Glu-OH.HCl
Formula:
C₅H₁₀ClNO₄ MW: 183.59 g/mol |
MDL Number:
MFCD00012619 CAS Number: 138-15-8 EINECS: 205-315-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...