Potassium-D-gluconate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Potassium Gluconate, Gluconic Acid Potassium Salt, D-Gluconic acid potassium salt, Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate

G0040-25G G0040-500G
TCG0040-025GEA 21.24 USD
TCG0040-025G TCG0040-500G
Potassium-D-gluconate ≥99.0% (by titrimetric analysis)
Potassium-D-gluconate

CAS Number: 299-27-4
MDL Number: MFCD00064211
Molecular Formula: C6H12O7
Molecular Weight: 234.25
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Specific rotation [a]20/D: 12 deg (C=2, H2O)


Product Categories: Bioscience, Sugar Acids, Glucose
Other Product Names / Synonyms: Gluconic Acid Potassium Salt

Formula: HOH₂C(CH(OH))₄COOK
MW: 234.25 g/mol
Melting Pt: 174 - 176 °C
Density: 1.73 g/cm³ (20 °C)
MDL Number: MFCD00064211
CAS Number: 299-27-4
EINECS: 206-074-2
Merck Index: 12,07796
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=448db2ef15757cd486fd60566656427157b85429&vid=448db2ef15757cd486fd60566656427157b85429&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR