D-Gluconic acid 45 - 50% in water (contains Gluconolactone)

Supplier: TCI America

Synonyms: Gluconic acid, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid

G0036-25G G0036-500G
TCG0036-025GEA 25.09 USD
TCG0036-025G TCG0036-500G
D-Gluconic acid 45 - 50% in water (contains Gluconolactone)
D-Gluconic acid

CAS Number: 526-95-4
MDL Number: MFCD00004240
Molecular Formula: C6H12O7
Molecular Weight: 196.16
Form: Clear Liquid
Color: Very Pale Yellow

Formula: C₆H₁₂O₇
MW: 196.16 g/mol
Density: 1.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004240
CAS Number: 526-95-4
EINECS: 208-401-4
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bbd7604fe3bb66d60773331201adf97bc08f582&vid=479b6a04954e160dd72aef34d3e419cfe8e4d30a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR