2-Fluoroadenosine ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (2R,3R,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

F0656-1G F0656-200MG
TCF0656-1GEA 287.97 USD
TCF0656-1G TCF0656-200MG
2-Fluoroadenosine ≥96.0% (by HPLC, titration analysis)
2-Fluoroadenosine

CAS Number: 146-78-1
MDL Number: MFCD00866394
Molecular Formula: C10H12FN5O4
Molecular Weight: 285.24
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Melting point (°C): 200
Specific rotation [a]20/D: -72 deg (C=0.1, EtOH)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: 6-Amino-2-fluoro-9-(beta-D-ribofuranosyl)purine

Formula: C₁₀H₁₂FN₅O₄
MW: 285.23 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00866394
CAS Number: 146-78-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92f3acb11c0b9c620db6f6e31676c9222c7410e3&vid=92f3acb11c0b9c620db6f6e31676c9222c7410e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR