3-Fluorothiophenol ≥97.0%
Supplier: TCI America
Synonyms:
3-Fluorobenzene-1-thiol, 1-Fluoro-3-mercaptobenzene, m-Fluorothiophenol, 3-Fluorobenzenethiol
F0422-25G
F0422-5G
TCF0422-25GEA
285
USD
TCF0422-25G
TCF0422-005G
3-Fluorothiophenol ≥97.0%
3-Fluorothiophenol
CAS Number: 2557-77-9
MDL Number: MFCD00040227
Molecular Formula: C6H5FS
Molecular Weight: 128.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 156
Flash Point (°C): 58
Specific Gravity (20/20): 1.20
Product Categories: Synthetic Organic Chemistry, Thiols, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluorothiophenol, 3-Mercaptofluorobenzene
Formula:
C₆H₅FS MW: 128.17 g/mol Boiling Pt: 156 °C Density: 1.20 (20 °C) Flash Pt: 58 °C |
MDL Number:
MFCD00040227 CAS Number: 2557-77-9 EINECS: 219-876-2 UN: 3071 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...