2-Fluorothiophenol ≥98.0%

Supplier: TCI America
Danger

Synonyms: 2-Fluorobenzenethiol, 1-Fluoro-2-mercaptobenzene, 2-Fluorobenzene-1-thiol, o-Fluorothiophenol

F0419-5G F0419-25G
TCF0419-005GEA 56 USD
TCF0419-005G TCF0419-25G
2-Fluorothiophenol ≥98.0%
2-Fluorothiophenol

CAS Number: 2557-78-0
MDL Number: MFCD00041419
Molecular Formula: C6H5FS
Molecular Weight: 128.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Specific Gravity (20/20): 1.20


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluorothiophenol, 2-Mercaptofluorobenzene

Formula: C₆H₅FS
MW: 128.17 g/mol
Boiling Pt: 164 °C
Density: 1.20 (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00041419
CAS Number: 2557-78-0
UN: 3071
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ad1fe8e974cd4f1d50c0bf7237c040d93fae4ae&vid=c079f549d933081047ec42051d243ee4a827d40f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR