4-Fluoro-o-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Amino-5-fluorotoluene, 4-Fluoro-2-methylaniline

F0398-25G F0398-5G
TCF0398-25GEA 103 USD
TCF0398-25G TCF0398-005G
4-Fluoro-o-toluidine ≥98.0% (by GC, titration analysis)
4-Fluoro-o-toluidine

CAS Number: 452-71-1
MDL Number: MFCD00007832
Molecular Formula: C7H8FN
Molecular Weight: 125.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 88
Freezing point (°C): 14
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 2-Amino-5-fluorotoluene, 4-Fluoro-o-toluidine

Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 102 °C
Melting Pt: 14 °C
Density: 1.12 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00007832
CAS Number: 452-71-1
EINECS: 207-208-2
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37c1810733344a0243aa16ccadd6c5f0b9d2abe3&vid=37c1810733344a0243aa16ccadd6c5f0b9d2abe3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR