4-Fluorothiophenol ≥98.0%
Supplier: TCI America
|
Danger
|
CAS Number: 371-42-6
MDL Number: MFCD00004846
Molecular Formula: C6H5FS
Molecular Weight: 128.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Flash Point (°C): 54
Specific Gravity (20/20): 1.20
Product Categories: Synthetic Organic Chemistry, Thiols, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluorothiophenol, 4-Mercaptofluorobenzene
Formula:
C₆H₅FS Boiling Pt: 168 °C Density: 1.20 (20 °C) Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004846 CAS Number: 371-42-6 EINECS: 206-737-6 UN: 3071 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...