2-Fluoro-4-nitrophenol ≥97.0%
Supplier: TCI America
F0381-5G
F0381-25G
TCF0381-005GEA
18.14
USD
TCF0381-005G
TCF0381-025G
2-Fluoro-4-nitrophenol ≥97.0%
2-Fluoro-4-nitrophenol
CAS Number: 403-19-0
MDL Number: MFCD00051970
Molecular Formula: C6H4FNO3
Molecular Weight: 157.10
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 120
Formula:
C₆H₄FNO₃ Melting Pt: 120 °C |
MDL Number:
MFCD00051970 CAS Number: 403-19-0 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...