4-Fluoro-2-nitrophenol ≥98.0%
Supplier: TCI America
F0367-5G
F0367-25G
TCF0367-005GEA
35
USD
TCF0367-005G
TCF0367-25G
4-Fluoro-2-nitrophenol ≥98.0%
4-Fluoro-2-nitrophenol
CAS Number: 394-33-2
MDL Number: MFCD00042528
Molecular Formula: C6H4FNO3
Molecular Weight: 157.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 75
Formula:
C₆H₄FNO₃ Melting Pt: 75 °C |
MDL Number:
MFCD00042528 CAS Number: 394-33-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...