3-Fluoro-4-nitrophenol ≥98.0%
Supplier: TCI America
|
Warning
|
F0348-25G
F0348-5G
TCF0348-25GEA
851.37
USD
TCF0348-25G
TCF0348-005G
3-Fluoro-4-nitrophenol ≥98.0%
3-Fluoro-4-nitrophenol
CAS Number: 394-41-2
MDL Number: MFCD00041251
Molecular Formula: C6H4FNO3
Molecular Weight: 157.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 92
Formula:
C₆H₄FNO₃ Melting Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041251 CAS Number: 394-41-2 EINECS: 206-895-6 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...