5-Fluorocytosine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one
CAS Number: 2022-85-7
MDL Number: MFCD00006035
Molecular Formula: C4H4FN3O
Molecular Weight: 129.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Antifungal Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleobases and Their Analogs
Other Product Names / Synonyms: 4-Amino-5-fluoro-2-hydroxypyrimidine, Flucytosine
Formula:
C₄H₄FN₃O MW: 129.09 g/mol Boiling Pt: 298 °C (1013 hPa) Melting Pt: 165 °C |
MDL Number:
MFCD00006035 CAS Number: 2022-85-7 EINECS: 217-968-7 Merck Index: 13,04151 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...